Learn More
Enzo Life Sciences N-Methyl-D-aspartic acid (50mg). CAS: 6384-92-5
Supplier: Enzo Life Sciences ALX105009M050
Selective ionotropic NMDA receptor agonist. Excitatory amino acid. Alternative name: NMDA. Formula: C5H9NO4. MW: 147.1. Purity: ≥98% (TLC). Solubility: Soluble in dilute aqueous base; slightly soluble in water or 100% ethanol. Long Term Storage: Ambient.