Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-D-prolinol, 99%, Thermo Scientific™

Catalog No. AAL1626803
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 83435-58-9
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
MDL Number MFCD00040580
InChI Key BFFLLBPMZCIGRM-MRVPVSSYSA-N
Synonym boc-d-prolinol, n-boc-d-prolinol, r-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, r-+-1-boc-2-pyrrolidinemethanol, 1-boc-d-prolinol, boc-d-pro-ol, n-tert-butoxycarbonyl-d-prolinol, n-t-boc-d-prolinol, tert-butyl 2r-2-hydroxymethyl pyrrolidine-1-carboxylate
PubChem CID 688279
IUPAC Name tert-butyl (2R)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1CO

Specifications

Melting Point 59°C to 62°C
Quantity 1 g
Optical Rotation +54° (c=5 in Methanol)
Formula Weight 201.27
Percent Purity 99%
Chemical Name or Material N-Boc-D-prolinol
TSCA No
Recommended Storage Keep cold
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.