Learn More
Enzo Life Sciences N-Arachidonylglycine (5mg). CAS: 179113-91-8
SDP
Supplier:  Enzo Life Sciences BMLFA0290005
Encompass_Preferred
Endogenous anandamide-like compound. Lacks affinity for CB1 receptors (Ki>10µM), TRPV1 (EC50>10µM) and anandamide uptake (IC50>50µM), but inhibits fatty acid amide hydrolase (FAAH) (IC50=8.5µM-50µM, depending on cell type and species). Displays anti-inflammatory and analgesic activity. Identified as an insulin secretagogue in a primary β-cell-based functional assay. Represents a new class of "lipoamino acids". Purity: ≥98% (TLC). Solubility: Soluble in DMSO or 100% ethanol. 2mg/ml soluble in PBS (pH 7.2). Long Term Storage: -20°C.
Catalog No. 50-201-0812
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.