Learn More
Enzo Life Sciences Mycophenolate mofetIL-(100mg). CAS: 128794-94-5
SDP
Supplier:  Enzo Life Sciences BMLIM1010100
Encompass_Preferred
Mycophenolate mofetil (CellCept) is an immunosuppressant prodrug of mycophenolic acid, a fermentation product of Penicillium brevicompactum. Immunosuppression occurs via inhibition of type II inosine-5’-monophosphate dehydrogenase, depleting guanosine preferentially in T and B lymphocytes. In addition, mycophenolic acid displays anti-inflammatory activity by preventing NO production specifically from iNOS and by blocking glycosylation and expression of adhesion molecules and therefore recruitment of lymphocytes and monocytes to sites of inflammation. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml, warm). Long Term Storage: Ambient.
Catalog No. 50-201-1044
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.