Learn More
Proteintech Group Inc Myc-Trap Magnetic Agarose
Supplier: Proteintech Group Inc YTMA20
The ChromoTek Myc-Trap Magnetic Agarose is an affinity resin for IP of Myc-fusion proteins. It consists of a Myc Nanobody/VHH coupled to magnetic agarose beads.
This item is not returnable.
View return policy