Learn More
Proteintech Group Inc Myc-Trap Agarose
SDP
Supplier:  Proteintech Group Inc YTA10
Encompass
Myc-Trap Agarose is an affinity reagent for IP and purification of Myc-tagged proteins. It consists of a Myc Nanobody/ VHH coupled to agarose beads.
Catalog No. 50-225-8935
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.