Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Multimedia Lessons for Interactive Whiteboards: Earth Science
SDPSCI_ED

The perfect complement to any earth science curriculum — use on the whiteboard for the entire class or on a computer for an individual student.

Supplier:  NewPath Learning 546301

 View more versions of this product

Catalog No. S02706


Description

Description

Key Features:

  • Virtual Lab Investigation: Allows students to experiment, perform simulations and draw conclusions
  • Custom Tool Bar: Allows students to draw, write and add arrows and text blocks to customize the lessons by adding details to any screen
Modules:
  • Lesson Preparation: Consists of a series of narrated, visual presentations, each featuring highly detailed graphics and illuminating animations
  • Interactive Activities: Includes a variety of interactive activities, a virtual lab investigation and a Lab Activity Guide; allows students to learn concepts, apply knowledge to other modules and develop skills to perform the virtual lab investigation
  • Visual Resources: Customize lessons using additional images, arrows, text and callouts provided in the Custom Tool Bar
  • Vocabulary: Illustrated vocabulary; show or hide definitions or play audio version
  • Assessment: Use in Quiz Mode, Practice Mode or Play Mode
  • Lesson Topics
    Identifying Minerals - What is a Mineral?, Properties of Minerals, Identifying Minerals, How Do Minerals Form?, Mineral Resources, Uses of Minerals; Virtual Lab Investigation: Identifying Minerals
    Rocks - Classifying Rocks, Igneous Rocks, Sedimentary Rocks, Metamorphic Rocks, Coral Reefs, The Rock Cycle; Virtual Lab Investigation: Identifying Rocks
    Plate Tectonics - Earth's Interior, Types of Heat Transfer, Continental Drift, Sea-Floor Spreading, Theory of Plate Tectonics; Virtual Lab Investigation: Plate Tectonics Simulation
    Earthquakes - Types of Stress, Kinds of Faults, How Earth's Surface Changes, Types of Seismic Waves, Measuring Earthquakes, Earthquake Damage and Safety; Virtual Lab Investigation: Modeling Earthquakes
    Volcanoes - Volcanoes and Plate Boundaries, Properties of Magma, Volcanic Eruptions, Life Cycle of a Volcano, Volcanic Landforms, Hot Springs and Geysers; Virtual Lab Investigation: Investigating Volcanoes
    Earth's Surface - Landforms, Topographic Maps, Weathering & Erosion, Mechanical Weathering, Chemical Weathering, Cycle of Erosion and Deposition, Soil; Virtual Lab Investigation: Effect of Chemical Weathering on Rocks
    Earth's Atmosphere and Weather - The Atmosphere, Weather Factors, Clouds, Weather Patterns, Predicting the Weather; Virtual Lab Investigation: Forecasting the Weather
    Earth's Climate - What is Climate?, Causes of Climate, Climate Regions/Types of Climate, Causes of Climate Change, Short-Term Climate Change, Global Warming, Ozone Depletion; Virtual Lab Investigation: Investigating Climate Change
    Our Solar System - Formation of the Solar System, Geocentric and Heliocentric Systems, The Sun, The Inner Planets, The Outer Planets, Comets, Asteroids and Meteors; Virtual Lab Investigation: Exploring Our Solar System
    Sun-Earth-Moon System - How Earth Moves; Seasons on Earth; Gravity and Motion; Phases, Eclipses and Tides; Earth's Moon; Missions to the Moon; Virtual Lab Investigation: Simulating the Sun-Earth-Moon System

    Order Info

    Lessons available with single-user or site license. Single-user license can be installed and used on a single computer/Interactive Whiteboard. Site license permits access to any computer/Interactive Whiteboard within a single school building.

    Certifications

    Accredited by SMART™ and Promethean™ for use on their Interactive Whiteboards.

Specifications

Specifications

6
Minerals Multimedia Lesson
Earth Science
5 Modules, digital inquiry based student activity guide
Single User
CD-ROM
8
Interactive whiteboard or computer
Minerals Multimedia Lesson
Smart™, Promethean™
Multisensory learning, perfect resource for interactive whiteboards and the lessons are also ideal for individual student instruction on any computer
Windows™, Mac™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.