Learn More
Andwin Scientific MULTIFLEX PIPET TIPS 800/CS
SDP
Supplier:  Andwin Scientific 13790
Encompass_Preferred
The MltiFlex Round gel loading tips have a volume capacity of 200 l (1 - 200 l). This product has graduation lines at 5 l. The tips are nonsterile and are sold in a case of 800.
Catalog No. NC0798279
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.