Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTJ1 Polyclonal Antibody, Bethyl Laboratories

Description
The recommended shelf life for this product is 1 year from date of receipt.
May modulate protein synthesis.Specifications
MTJ1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q61712, Q96KC8 | |
DNAJC1 | |
Between 475 and 525 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
DNAJC1 | |
DNAJL1, ERdj1, HTJ1, MTJ1, DnaJ-like protein, dnaJ homolog subfamily C member 1, dnaJ protein homolog MTJ1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13418, 64215 | |
4° C | |
Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?