Learn More
Regis Technologies Inc GC Regeant, Regis, MTBSTFA + 1% TBDMCS (25 GM)
SDP
Supplier:  Regis Technologies Inc 1270143200
Encompass_Preferred
MtBSTFA is a Silylation Reagent. MtBSTFA replaces active hydrogens to form t-BDMS derivatives. Derivatization is usually complete upon dissolution with this exceptionally strong, yet mild silylating reagent. MtBSTFA derivatives are 104 times more stable to hydrolysis than their corresponding TMS derivatives and produce easily interpreted mass spectra for GC/MS. Addition of t-BDMCS catalyzes reactions of hindered alcohols and amines.
Catalog No. 50-442-892
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.