Learn More
Eurofins Discoverx αMSH, 1 mg. (92-1172)
Supplier:  Eurofins Discoverx 921172
Encompass_Preferred
α-Melanocyte stimulating hormone (α-MSH) is a pituitary hormone that plays a key role in appetite control and the skins UV protection and development of pigmentation. Also has effects on energy homeostasis, sexual activity, and as an anti-inflammatory. Potent agonist of the melanocortin receptors MC1, MC3, MC4, and MC5 (Ki=0.12, 31, 660, and 5700 nM, respectively), but not MC2. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9700
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.