Learn More
G & P Biosciences MSE MSLN/MESOTHLN LENTIV, 1ML
Supplier:  G & P Biosciences LTV1588
Encompass_Preferred
Mouse MSLN/Mesothelin Lentivirus, Full-length Gene in Lentivector, Pre-packaged lentiviral Particles, 1ml, Puromycin marker.
Catalog No. NC2253989
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.