Learn More
Innovative Research Inc Ms tPA Rcmb Non-Enz/Cleav 1mg
Supplier: Innovative Research Inc IMSTPARCNENC1MG
Mouse tPA Recombinant Non-Enzymatic Non-Cleavable 1mg - Mouse tPA Recombinant Non-Enzymatic Non-Cleavable is recombinantly produced in insect cells and purified using affinity chromatography. This is a frozen liquid buffered in 0.4M HEPES; 0.1M NaCl; pH 7.4 and a purity of >95%. This product is useful as a positive control for ELISA and western blot and for a variety of in vitro applications including cell differentiation, cell signaling, clotting assays, fibrinolysis, plasminogen activation, protein-protein interactions, receptor binding, thromboelastography, and thrombolysis. Mouse tPA with a double mutation that is permanently single chain and lacks catalytic activity. The non-catalytic mutation site is S481A on the mature protein and S510A on the complete mRNA sequence (UniProtKB locus TPA_MOUSE, accession P11214). The non-cleavable mutation site is 279E on the mature protein and R308E on the complete mRNA sequence (UniProtKB locus TPA_MOUSE, accession P11214).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.