Learn More
Innovative Research Inc Ms tPA Rcmb Act 100ug
Supplier:  Innovative Research Inc IMSTPAAR100UG
Encompass_Preferred
Mouse tPA Recombinant Active 100ug - Mouse tPA Active Recombinant from Innovative Research has been recombinantly produced in insect cells and purified using affinity chromatography. This is a frozen liquid buffered in 0.4M HEPES; 0.1M NaCl; pH 7.4 and a purity of >95%. This product is useful as a positive control for ELISA and western blot and for a variety of in vitro applications including cell differentiation, cell signaling, clotting assays, fibrinolysis, plasminogen activation, protein-protein interactions, receptor binding, thromboelastography, and thrombolysis. >95% active. More Details Source Insect Cell Culture Purification Affinity purified Purity >95% Storage Conditions -70C
Catalog No. 50-274-4134
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.