Learn More
Innovative Research Inc Ms Protse Nxn 1 Ntv Rcmb 100ug
Supplier:  Innovative Research Inc IMSPN1NR100UG
Encompass_Preferred
Mouse Protease Nexin 1 Native Recombinant 100ug - Mouse Protease Nexin 1 Native Recombinant from Innovative Research has been recombinantly produced in insect cell culture and purified by affinity chromatography. This is a Frozen liquid buffered in 0.01M Sodium Phosphate; 0.3M NaCl; pH 7.0 and a purity of 95% as determined by SDS-PAGE. More Details Species Mouse Target Protease Nexin 1 Purification Method Affinity purified Purity 95% as determined by SDS-PAGE Analysis Source Insect cell culture Storage Conditions -70C Additional Information Active and fully complexable with tissue-type plasminogen activator. Inhibits serine proteases such as thrombin, tPA and uPA. This product can be used as a control in applications like ELISA and Western Blot. It can be used for a wide range of in vitro applications, including fibrinolysis, hemostasis, inhibition, protein-protein interactions, and more.
Catalog No. 50-274-4139
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.