Learn More
Innovative Research Inc Ms Prorenin Pur 8X-Hist 100ug
Supplier:  Innovative Research Inc IMSPREN8XHIS100UG
Encompass_Preferred
Mouse Prorenin Purified 8X-Histidine 100ug - Mouse Prorenin Purified 8X-Histidine from Innovative Research has been recombinantly produced in HEK cells and purified using chelated metal affinity chromatography. This is a frozen liquid buffered in 50 mM Tris; pH 8.0 and a purity of >95%. This product is useful as a positive control for ELISA and western blot and for a variety of in vitro and in vivo applications including cell signaling, hypertension studies, protein-protein interactions, receptor binding. Resistant to activation to renin by trypsin digestion. More Details Source Human Embryonic Kidney Cells Purification Chromatographically purified Purity >95% Storage Conditions -70C
Catalog No. 50-274-4138
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.