Learn More
Innovative Research Inc Ms CD1 RBCs 100ml
Supplier:  Innovative Research Inc IMSCD1RBC100ML
Encompass_Preferred
Mouse CD1 Red Blood Cells 100ml - Mouse CD1 Red Blood Cells from Innovative Research are manufactured using whole blood collections from type CD1 mice. The whole blood is centrifuged to separate the platelet-rich plasma, buffy coat layer, and leukocytes. Red blood cells are perishable and are collected and processed upon receipt of order.
Catalog No. 50-274-4172
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.