Learn More
Enzo Life Sciences MPEP . hydrochloride (25mg). CAS: 96206-92-7
SDP
Supplier:  Enzo Life Sciences ALX550368M025
Encompass_Preferred
Potent and selective antagonist for mGluR5 receptor. Systemically active in vivo. Alternative name: 6-Methyl-2-(phenylethynyl)-pyridine . HCl. Purity: ≥99%. Solubility: Soluble in water, DMSO or methanol. Long Term Storage: +4°C.
Catalog No. 50-200-9548
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.