Learn More
World Precision Instrument Mouse Stereotaxic Anesthesia Mask with Bite Bar
Supplier: World Precision Instrument 505385
WPI's 505226 is compatible for use with the 505313 mouse stereotaxic instruments including: Mouse and Neonatal Rat Adaptor (Product Number 505243), Just for Mouse Stereotaxic Series (Product Number 505313) and Just for Mice Stereotaxic (Product Number 505243) Series.
Adjustable snout clamp provides secure positioning for direct gas delivery. Inlet/outlet and outlet tubing connections allow for continuous flow of anesthesic gas and active gas scavenging from gas scavenger as shown in the image below.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.