Learn More
STEMCELL Technologies Mouse Recombinant SCF, Size: 1000 µg
SDP
Supplier:  STEMCELL Technologies 78064.2
Encompass_Preferred
Use stem cell factor (SCF) to promote cell survival, proliferation, differentiation, adhesion, and functional activation of cells at multiple levels of the hematopoietic hierarchy. SCF is an early-acting cytokine that activates PI3K, JAK/STAT, and MAPK pathways upon binding to its receptor. Together with other cytokines like thrombopoietin and Flt3/Flk-2 Ligand, SCF is used to promote expansion of primitive hematopoietic stem cells and multipotent progenitor cells in culture (Martin et al. Cell, 1990; Kent et al. Clin Cancer Res, 2008.). In combination with various growth factors and erythropoietin, SCF increases proliferation and differentiation of myeloid and erythroid progenitor cells and a subset of lymphoid progenitor cells (Broudy VC. Blood, 1997). Mouse recombinant SCF from STEMCELL comes lyophilized with ≥ 95% purity; EC50 ≤ 20 ng/mL; endotoxin levels verified ≤ 1 EU/μg protein. Alternative names: Kit ligand, Mast cell growth factor, Steel factor, Stem cell factor.
Catalog No. 50-197-6388
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.