Learn More
Proteintech Group Inc Mouse IL-1ra/IL-1F3 ELISA Kit
SDP
Supplier:  Proteintech Group Inc KE1005296T
Encompass_Preferred
KE10052 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The IL-1ra ELISA kit is to be used to detect and quantify protein levels of endogenous IL-1ra. The assay recognizes mouse IL-1ra. An antibody specific for IL-1ra has been pre-coated onto the microwells. The IL-1ra protein in samples is captured by the coated antibody after incubation. Following extensive washing, another antibody of biotinylated specific for IL-1ra is added to detect the captured IL-1ra protein. For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent. Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.
Catalog No. 50-225-8459
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.