Learn More
Sino Biological Mouse IgG PAb 20UG 50323-T16-H-20
Supplier:  Sino Biological 50323T16H20
Encompass_Preferred
Produced in rabbits immunized with purified mouse IgG (heavy and light chains). This polyclonal antibody was purified by antigen affinity, and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-240-7814
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.