Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Mouse IGF-I Recombinant Protein, PeproTech®
E. coli Recombinant Protein
$89.50 - $1552.00
Specifications
Accession Number | P05017 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 16000 |
Molecular Weight (g/mol) | 7.6 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
25019100UG
|
Gibco™
25019100UG |
100 μg |
Each for $311.00
|
|
|||||
250-19-10UG
|
Gibco™
2501910UG |
10 μg |
Each for $89.50
|
|
|||||
250-19-1MG
|
Gibco™
250191MG |
1 mg |
Each for $1,552.00
|
|
|||||
25019250UG
|
Gibco™
25019250UG |
250 μg |
Each for $544.00
|
|
|||||
25019500UG
|
Gibco™
25019500UG |
500 μg |
Each for $932.00
|
|
|||||
250-19-50UG
|
Gibco™
2501950UG |
50 μg |
Each for $217.00
|
|
|||||
Description
IGF1 (Insulin-like growth factor-1) is structurally and functionally related to insulin but has a much higher growth-promoting activity. A variety of cellular responses are induced by IGF1, including cell proliferation, differentiation, migration, and survival. Further, IGF1 is a polypeptide growth factor that stimulates the proliferation of a wide range of cell types in muscle, bone, and cartilage tissue. IGF1 stimulates glucose transport in rat bone-derived osteoblastic (PyMS) cells and is effective at much lower concentrations than insulin, not only regarding glycogen and DNA synthesis but also with regards to enhancing glucose uptake. In circulation, IGFs are predominantly bound to binding proteins (IGFBPs) which prolong the half-life of the IGFs and play a role in delivering them to target tissues. IGF-I is known as one of the most potent activators of the AKT signaling pathway which is known to be a stimulator of proliferation and an inhibitor of programmed cell death. Moreover, mature human IGF-I is 100% homologous with bovine and porcine proteins. Low levels of IGF1 have been linked to Alzheimer's disease. IGF1 is processed from a precursor, bound by a specific receptor, and secreted. Defects in the IGF1 gene are a cause of insulin-like growth factor 1 deficiency and several transcript variants encoding different isoforms have been found.Specifications
P05017 | |
protein with no preservative | |
7.6 | |
E. coli | |
RUO | |
C730016P09Rik; class 1 insulin-like growth factor I preproprotein; class 2 insulin-like growth factor I preproprotein; H-IGF-1; IBP1; IGF; IGF1; Igf-1; IGF1A; IGFI; Igf-I; IGFIa; IGF-IA; IGF-IB; insulin growth factor-1; Insulin like growth factor; insulin like growth factor 1; insulin-like growth factor 1; insulin-like growth factor 1 (somatomedin C); insulin-like growth factor I; insulin-like growth factor I (somatomedin C); insulin-like growth factor IB; mechano growth factor; MGF; M-IGF-1; OTTHUMP00000195084; prepro-IGF-I; prepro-insulin-like growth factor I; R-IGF-1; somatomedin; Somatomedin C; Somatomedin-C | |
Mouse | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
16000 | |
Mouse IGF-I | |
-20°C | |
<1 EU/μg | |
IGF1 | |
Unconjugated | |
GPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKAA | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title