Learn More
PBL Assay Science Mouse Interferon Beta, mammalian (IFN-Beta, IFNb) [1e5 (100,000) units/vial]
Supplier: PBL Assay Science 124051
Recombinant protein Mouse Interferon Beta, mammalian is manufactured by PBL Assay Science (Catalog #12405-1) for research use only (RUO) and packaged in 1e5 (100,000) units/vial. This cytokine exhibits 10-fold higher specific activity than E.coli-derived Mu IFN-Beta and has an authentic sequence without a tag.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.