Learn More
ACROBiosystems Anti-His Tag Antibody, Human IgG1-coupled Magnetic Beads (recommended for MPCLIA)
Supplier:  ACROBiosystems MPCA00710MG
Encompass_Preferred
The Anti-His Tag-coupled Magnetic Beads are 2.8 µm superparamagnetic particles covalently coupled to a highly affinity Monoclonal Anti-His Tag antibody. The beads can be used to capture the proteins containing C-terminal, N-terminal, and internal His tagged fusion in Chemiluminescence procedures. The Monoclonal Anti-His Tag antibody is a chimeric monoclonal antibody expressed from HEK293 cells, which combines the variable region of a mouse monoclonal antibody with human IgG1 constant domain. The Anti-His Tag-coupled Magnetic Beads is easy to capture his Tagged protein or other molecules, this ready to use products could greatly save your protein coupling time and hassle, and help us get the best performance and highly reproducible results.
Catalog No. 50-233-1245
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.