Learn More
Sino Biological Mouse CSF2 ELISA Kit 1UN (Sino Biological)
Supplier:  Sino Biological KIT510481
Encompass_Preferred
For the quantitative determination of Mouse GM-CSF/CSF2 concentration in serum and cell culture supernates.
Catalog No. 50-237-3479
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.