Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Mouse CCL22 (MDC) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
$87.00 - $3240.00
Specifications
Accession Number | O88430 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 20299 |
Molecular Weight (g/mol) | 7.8 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
25023100UG
|
Gibco™
25023100UG |
100 μg |
Each for $648.00
|
|
|||||
250-23-1MG
|
Gibco™
250231MG |
1 mg |
Each for $3,240.00
|
|
|||||
250-23-20UG
|
Gibco™
2502320UG |
20 μg |
Each for $211.00
|
|
|||||
25023250UG
|
Gibco™
25023250UG |
250 μg |
Each for $1,134.00
|
|
|||||
25023500UG
|
Gibco™
25023500UG |
500 μg |
Each for $1,944.00
|
|
|||||
250-23-5UG
|
Gibco™
250235UG |
5 μg |
Each for $87.00
|
|
|||||
Description
CCL22 may play a role in the trafficking of activated/effector T-lymphocytes to inflammatory sites and other aspects of activated T-lymphocyte physiology. Also, it is a chemotactic for monocytes. dendritic cells and natural killer cells. CCL22 is a mild chemoattractant for primary activated T-lymphocytes and a potent chemoattractant for chronically activated T-lymphocytes but has no chemattractant activity for neutrophils, eosinophils, and resting T-lymphocytes. Binds to CCR4.Specifications
O88430 | |
protein with no preservative | |
7.8 | |
E. coli | |
RUO | |
A-152E5.1; Abcd1; ABCD-1; Activated B and dendritic cell-derived; AW060611; CC chemokine ABCD-1; CC chemokine STCP-1; C-C motif chemokine 22; C-C motif chemokine ligand 22; Ccl22; chemokine (C-C motif) ligand 22; DC/B-CK; DCBCK; dendritic cell and B cell derived chemokine; macrophage-derived chemokine; MDC; MDC(1-69); MDC(3-69); MDC(5-69); MDC(7-69); MGC108943; MGC34554; RP24-69A20.1; Scya22; small inducible cytokine A22; SMALL INDUCIBLE CYTOKINE A22 PRECURSOR (CC CHEMOKINE ABCD-1) (ACTIVATED B AND DENDRITIC CELL-DERIVED); small inducible cytokine subfamily A (Cys-Cys) member 22; small inducible cytokine subfamily A (Cys-Cys), member 22; small inducible cytokine subfamily A, member 22; small inducible cytokine subfamily A22; small-inducible cytokine A22; STCP-1; stimulated T cell chemotactic protein 1; stimulated T-cell chemotactic protein 1 | |
Mouse | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
20299 | |
Mouse CCL22 (MDC) | |
-20°C | |
<1 EU/μg | |
CCL22 | |
Unconjugated | |
GPYGANVEDS ICCQDYIRHP LPSRLVKEFF WTSKSCRKPG VVLITVKNRD ICADPRQVWV KKLLHKLS | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title