Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Mouse CCL19 (MIP-3 beta) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
$87.00 - $3240.00
Specifications
Accession Number | O70460 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 24047 |
Molecular Weight (g/mol) | 9.2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
25027B100UG
|
Gibco™
25027B100UG |
100 μg |
Each for $648.00
|
|
|||||
250-27B-1MG
|
Gibco™
25027B1MG |
1 mg |
Each for $3,240.00
|
|
|||||
25027B20UG
|
Gibco™
25027B20UG |
20 μg |
Each for $211.00
|
|
|||||
25027B250UG
|
Gibco™
25027B250UG |
250 μg |
Each for $1,134.00
|
|
|||||
25027B500UG
|
Gibco™
25027B500UG |
500 μg |
Each for $1,944.00
|
|
|||||
250-27B-5UG
|
Gibco™
25027B5UG |
5 μg |
Each for $87.00
|
|
|||||
Description
CCL19, also known as MIP-3 beta (Macrophage Inflammatory Protein 3 beta), is a member of the CC- subfamily of chemokines. It is most closely related to CCL21, with which it shares 32% amino acid sequence homology. CCL19 and CCL21 are expressed mainly by stromal cells in the T cell-rich zones of lymph nodes. They are critical mediators of the homeostatic trafficking of naive T cells and activated dendritic cells into the secondary lymphoid organs. CCL19 and CCL21 also play a role in T cell priming and activation, as well as the recruitment of lymphocytes to inflamed tissue. Both proteins signal via the G protein-coupled receptor, CCR7, which is expressed on T cells and mature dendritic cells. Although CCL19 and CCL21 both exhibit the same affinity for CCR7, only the binding of CCL19 results in the desensitization and internalization of the receptor.Specifications
O70460 | |
protein with no preservative | |
9.2 | |
E. coli | |
RUO | |
beta chemokine exodus-3; beta-chemokine exodus-3; CC chemokine ligand 19; C-C motif chemokine; C-C motif chemokine 19; C-C motif chemokine ligand 19; Ccl19; chemokine (C-C motif) ligand 19; chemokine CCL19; CK beta-11; CKb11; EBI1 ligand chemokine; EBI-1 ligand chemokine; EBI1-ligand chemokine; EBV-induced molecule 1 ligand chemokine; ELC; Epstein-Barr virus-induced molecule 1 ligand chemokine; exodus-3; macrophage inflammatory protein 3 beta; macrophage inflammatory protein 3-beta; MIP3B; MIP-3 b; MIP-3-beta; M-MIP-3-beta; SCYA19; small inducible cytokine A19; small inducible cytokine subfamily A (Cys-Cys), member 19; small-inducible cytokine A19 | |
Mouse | |
Recombinant | |
Lyophilized |
Functional Assay | |
24047 | |
Mouse CCL19 (MIP-3 beta) | |
-20°C | |
<1 EU/μg | |
Ccl19 | |
Unconjugated | |
GANDAEDCCL SVTQRPIPGN IVKAFRYLLN EDGCRVPAVV FTTLRGYQLC APPDQPWVDR IIRRLKKSSA KNKGNSTRRS PVS | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title