Learn More
IgG2b Mouse anti-Rat, eFluor 660, eBioscience™ Secondary Antibody, Invitrogen™
Mouse Monoclonal Antibody
$109.00 - $236.00
Specifications
Antigen | Rat IgG2b |
---|---|
Clone | R2B-7C3 |
Applications | Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Conjugate | eFluor 660 |
Catalog Number | Mfr. No. | dimQuantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | dimQuantity | Price | Quantity & Availability | |||||
50-481-582
|
LIFE TECHNOLOGIES
50481582 |
Each for $236.00
|
|
||||||
50-481-580
|
LIFE TECHNOLOGIES
50481580 |
Each for $109.00
|
|
||||||
Description
The MAb R2B-7C3 recognizes Rat IgG2b antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other rat isotypes nor does it crossreact to mouse IgG2b or any mouse isotype antibodies.
Rat IgG2b Monoclonal antibody specifically detects Rat IgG2b in Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)Specifications
Rat IgG2b | |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen) | |
eFluor 660 | |
Mouse | |
Rat | |
IgG1 κ | |
Affinity chromatography |
R2B-7C3 | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.