Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Potassium Channel Kir4.1, Clone: 9C9.2,

Mouse Monoclonal Antibody

Supplier:  MilliporeSigma™ MABN864

Catalog No. MABN864


Explore all special offers
Add to Cart

Description

Description

Specifically detects Potassium Channel Kir4.1 clone: 9C9.2 in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

ATP-sensitive inward rectifier potassium channel 10 (UniProt P78508; also known as ATP-dependent inwardly rectifying potassium channel Kir4.1, Inward rectifier K+ channel Kir1.2, Potassium channel, inwardly rectifying subfamily J member 10) is encoded by the KCNJ10 (also known as SESAMES) gene (Gene ID 3766) in human. Inward rectifier K+ channels (Kirs) modulate cell membrane potential and are classified as K+ transport channels (Kir1.x, Kir4.x, Kir5.x, Kir7.x), classic Kir (Kir2.x), G-protein-gated Kirs, and ATP-sensitive Kirs. Kirs mediate diverse physiological functions of both excitable and nonexcitable epithelial cells, such as cell adhesion-migration, volume regulation, apoptosis, and proliferation. Kir4.1 is widely expressed in the kidney distal convoluted tubules and neural glia cells, its expression in the cochlea and retina is also reported. Kir4.1 modulates plasma K+ by favoring K+ exit, and inhibition of Kir4.1 results in membrane depolarization. KCNJ10 gene mutations are known causes of SeSAMES (seizures, sensorineural deafness, ataxia, mental retardation, and electrolyte imbalance syndrome). Human Kir4.1 is a 379-amino acid multi-pass membrane (a.a. 65-89, 115-126, 143-164) protein containg two small extraceullar regions (a.a. 90-114 and 134-142), a pore-forming domain (a.a. 134-142), and having both N- and C-terminal exposed intracellularly (a.a. 1-64 and a.a. 165-379).
Specifications

Specifications

Potassium Channel Kir4.1
9C9.2
P78508
KCNJ10; SESAMES
KLH-conjugated linear peptides corresponding to sequences from the first extracellular domain and the C-terminal cytoplasmic tail of human potassium channel Kir4.1.
Purified by Ion-Exchange Chromatography
RUO
Stable for one year at 2°C to 8°C from date of receipt.
Monoclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
Purified mouse monoclonal IgMκ antibody in PBS with 0.05% sodium azide.
Purified
Mouse
IgM, κ
100μg
Neuroscience
Primary
NP_002232
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only