Learn More
IRF6, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant IRF6.
Supplier: Abnova Corporation H00003664A01
Description
This gene encodes a member of the interferon regulatory transcription factor (IRF) family. Family members share a highly-conserved N-terminal helix-turn-helix DNA-binding domain and a less conserved C-terminal protein-binding domain. Mutations in this gene can cause van der Woude syndrome and popliteal pterygium syndrome. This protein is involved in palate formation. [provided by RefSeq
Sequence: PFEIYLCFGEEWPDGKPLERKLILVQVIPVVARMIYEMFSGDFTRSFDSGSVRLQISTPDIKDNIVAQLKQLYRILQTQESWQPMQPTPSMQLPPALPPQSpecifications
IRF6 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant IRF6. | |
IRF6 | |
LPS/OFC6/PIT/PPS/VWS | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_006147 | |
IRF6 | |
IRF6 (NP_006138, 368 a.a. ∽ 467 a.a) partial recombinant protein with GST tag. | |
RUO | |
3664 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
For Research Use Only