Learn More
Bethyl Laboratories, Inc Mouse Albumin ELISA Kit, 1.23 – 900 ng/ml
Supplier: Bethyl Laboratories, Inc E99134
Mouse Albumin ELISA Kit, 1 xpre-coated 96-well strip plate, standard, detection antibody, sample dilution buffer, HRP solution, TMB substrate, stop solution, 20X wash buffer, sealing tape