Learn More
New England Biolabs, Inc. Monarch RNA Cleanup Wash Buffer – 40 ml
SDP
Supplier:  New England Biolabs, Inc. T2042L
Encompass
The Monarch RNA Cleanup Wash Buffer, a component of the Monarch RNA Cleanup Kits, is used to wash RNA after binding to ensure the removal of contaminants such as enzyme, NTPs, and short RNA fragments. It is supplied as a 5X concentrate and requires dilution with ethanol (>=95%) before use. The product is supplied in 2 bottles, each with 20 ml of concentrated buffer.
Catalog No. 50-195-9392
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.