Learn More
Hach Company MolyVer« 3 Reagent Powder Pillows, 10 mL, pk/100
Supplier: Hach Company 2604499
For high range molybdenum determination by the Mercaptoacetic Acid method. Sample Size: 10 mL Hach Method 8036. Pack of 100 powder pillows.