Learn More
Hach Company MolyVer 2 Reagent Powder Pillows, 25 mL, pk/100
Supplier:  Hach Company 1414869
Encompass_Preferred
For determination of high range molybdenum by the mercaptoacetic acid method (Hach method 8036). Range: up to 40.0 mg/L Mo Sample Size: 25 mL. Pack of 100 powder pillows
Catalog No. 50-199-4935
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.