Learn More
Hach Company Molybdovanadate Reagent, 500 mL
Supplier:  Hach Company 2076049
Encompass_Preferred
For phosphate (phosphorus) determination by the Molybdovanadate Method: Hach method 8114. Range: up to 45.0 mg/L PO4 3-. 500 mL bottle. Use with a bottle-top dispenser for Rapid Liquid procedures.
Catalog No. 50-199-5239
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.