Learn More
Electron Microscopy Sciences Molybdenum Boat, 12 mm x 5 mm x 1 mm , 5/pk
Supplier: Electron Microscopy Sciences 73810
For disc aperture cleaning, vacuum evaporation. Thickness: 0.05 mm (0.002") Overall Length is 75 mm (3"). Trough Measures: 12 mm(1.2")long x 5 mm (3/16")wide x 1 mm deep