Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pro-Q™ Diamond Phosphoprotein Gel Destaining Solution
SDP

Pro-Q Diamond Phosphoprotein Gel Destaining Solution is a ready-to-use solution premixed for your convenience.

$99.75 - $325.00

Specifications

Content And Storage Store at room temperature.
Detection Location In-Gel Detection
Detection Method Fluorescence
Label or Dye Pro-Q™ Diamond
Product Line PRO-Q™
View More Specs

Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
P33310
SDP
Encompass
View Documents
Invitrogen™
P33310
Pro-Q™ Diamond Phosphoprotein Gel Destaining Solution, 1 L
Each for $99.75
Add to Cart
 
P33311
SDP
Encompass
View Documents
Invitrogen™
P33311
Pro-Q™ Diamond Phosphoprotein Gel Destaining Solution, 5 L
Each for $325.00
Add to Cart
 
Description

Description

Pro-Q Diamond Phosphoprotein Gel Destaining Solution is a ready-to-use solution premixed for your convenience. It is formulated for use with Pro-Q Diamond Phosphoprotein Gel Stain. Destaining is important for reducing the gel background signal and the signal from nonspecific staining.

Pro-Q Diamond Phosphoprotein Gel Destaining Solution is a ready-to-use solution premixed for your convenience. It is formulated for use with Pro-Q Diamond Phosphoprotein Gel Stain. Destaining is important for reducing the gel background signal and the signal from nonspecific staining.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Store at room temperature.
Fluorescence
PRO-Q™
Proteins (Phosphoproteins)
In-Gel Detection
Pro-Q™ Diamond
Room Temperature
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.