Learn More
Electron Microscopy Sciences ThermoSafe Model 600 Drug Test Kit Mailer
Supplier:  Electron Microscopy Sciences 6151400
Encompass_Preferred
All in one, easy to use drug test mailing system that meets postal and courier service regulations for non-hazardous clinical specimens. Model 600 kit consists of 90 ml specimen bottle, screw top cap and temperature indication label, pad of Absorbent Material, Press-Lock Plastic Ba, Model 601, Model 602, and Security Seal.
Catalog No. 50-337-97
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.