Learn More
Electron Microscopy Sciences U-tek Model 444 Mini Mailers U-tek Economy Packs
Supplier:  Electron Microscopy Sciences 6152144
Encompass_Preferred
Compact protection for temperature sensitive shipments, ideal for serums, tissues, and reagents. Accommodate tubes, vials, ampoules, small bottles, and tissue flasks with space for refrigerants. Insulating, thick wall polystyrene foam construction. Holds Model 440 Mini Mailer and Model 429 U-tek refrigerant packs -1 deg C (+30 deg F).
Catalog No. 50-338-21
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.