Learn More
Electron Microscopy Sciences U-tek Model 406J U-tek Ice Packets
Supplier:  Electron Microscopy Sciences 6153306
Encompass_Preferred
Cushion fragile shipments, won't dilute ice baths or damage samples. They remain frozen longer than regular ice without the melting mess. May be refrozen and reuse, withstands pressure of 60 lb per square inch for 5 minutes or longer. Refrigerant contains neutral pH distilled deionized water and proprietary materials certified by the US Food and Drug Administration as food grade chemicals. -1 degree C / +30 degree F. Individual Packets.
Catalog No. 50-339-26
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.