Learn More
Electron Microscopy Sciences ThermoSafe Model 341 LabMailer
Supplier: Electron Microscopy Sciences 6151541
Maximum breakage protection for known non-hazadous blood, serum, tissue, and other specimens. Accommodates VacuTainer Microtainer and other collection / transport tubes. Lightweight polystyrene foam construction saves postage. Reusable and recyclable. Holds 8 tubes, 113 x 18, 8 slides, 342 Mailing Sleeve.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.