Learn More
Proteintech Group Inc mNeonGreen-Trap Magnetic Agarose, kit
Supplier: Proteintech Group Inc NTMAK20
The ChromoTek mNeonGreen-Trap Magnetic Agarose is an affinity resin for IP of mNeonGreen-fusion proteins. It consists of a mNeonGreen Nanobody/ VHH coupled to magnetic agarose beads.
This item is not returnable.
View return policy