Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ ML 00253764 hydrochloride
Melanocortin MC4 receptor antagonist; brain penetrant
$370.50 - $1547.00
Specifications
Purity | >98% |
---|---|
Product Type | ML 00253764 hydrochloride |
Molecular Weight (g/mol) | 413.71 |
Inhibitors | ML 00253764 hydrochloride |
Description
Non-peptide, melanocortin MC4 receptor antagonist (IC50 values are 320, 810 and 2120 nM for MC4, MC3 and MC5 receptors respectively). Decreases cAMP accumulation in HEK-293 cells expressing the MC4 receptor. Increases food intake and reduces loss of lean body mass in tumor bearing mice. Brain penetrant.Specifications
>98% | |
ML 00253764 hydrochloride | |
413.71 | |
ML 00253764 hydrochloride |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title