Learn More
Anton Paar MKT 10 Portable handheld Millikelvin Thermometer
Supplier: Anton Paar 172072
MKT 10 is a lightweight and easily portable handheld Millikelvin Thermometer which measures temperatures with an accuracy of 10 mK (0.01°C). MKT 10 comes with a Pt-100 probe and a carrying case for safe transport. Ideal for different applications: from temperature measurement in the production process to different calibrations in the lab. Highly stable Pt 100 sensor with factory certificate.Recommended accessory Protection tube for MKT 10 Probe NC1818338Measuring Range from 0°C up to 100°CAccuracy: 0.01°C (10 mK)Resolution: 0.001°C (1 mK)Ambient temperature suitable for 0°C up to 35°C (highest accuracy 20°C up to 25°C)Power Supply 4x AA batteries or Power over Ethernet (PoE)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.