Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mirus Bio™ TransIT-TKO™ siRNA Transfection Reagent
Enables high efficiency siRNA delivery and knockdown of target gene expression in many cell types, including primary cells. siRNA and DNA co-transfection is feasible with TransIT-TKO. Transfections do not require medium changes and can be performed in serum-containing medium.
Supplier: Mirus Bio™ MIR 2150
Description
- Broad Spectrum siRNA Delivery: Utilize one transfection reagent and protocol for a wide variety of cells
- Low Cellular Toxicity: Maintain cell density and reduce experimental biases
- Reproducible Results: Obtain consistent, targeted gene knockdowns from day to day
- High Knockdown Efficiency: Achieve optimal gene silencing in a large percentage of cells to ensure experimental success
Specifications
Tube | |
Mammalian | |
Chemical Transfection | |
Transfect small interfering RNA for knock out of target gene expression, optimal gene silencing in a large percentage of cells |
Sterile | |
None (Promoterless) | |
1.5 mL | |
1.37 mg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Share a Content Correction