Learn More
Electron Microscopy Sciences miniLIGHT Set 2 1W
Supplier: Electron Microscopy Sciences 7166502
Constructed from the highest quality material and small as its name, this miniLight offers the most universal light source, handling many tasks that require a bright light but with a small foot print that fits into most locations.
The miniLight's main component is the miniLamp. This is a small but powerful light source. It shines very bright on a small area. It has a low dissipation loss with no hot part. Long life because the lifetime of a LED is 50,000 hours. Little power needed (1watt). Adjustable intensity and CE compatible. Light source 28 deg , Gooseneck, Power Supply, and Steel Base.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.