Learn More
Electron Microscopy Sciences miniLIGHT Complete Set 1W
Supplier:  Electron Microscopy Sciences 7166505
Encompass_Preferred
Constructed from the highest quality material and small as its name, this miniLight offers the most universal light source, handling many tasks that require a bright light but with a small foot print that fits into most locations.
The miniLight's main component is the miniLamp. This is a small but powerful light source. It shines very bright on a small area. It has a low dissipation loss with no hot part. Long life because the lifetime of a LED is 50,000 hours. Little power needed (1watt). Adjustable intensity and CE compatible. Includes: all three Light Sources 15, 28 & 42 deg , Gooseneck, Mounting Rods, Joint, Power Supply, Magnet Base, and Steel Base.
Catalog No. 50-346-41
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.