Learn More
Electron Microscopy Sciences Mini Rotator, 220 V
Supplier: Electron Microscopy Sciences 6510020
The mini-rotator is a small, compact, rotator for micro centrifuge tubes and micro test tubes. It is heavy duty and has a permanent-magnet motor which maintains constant speed between 2-80 rpm. Mixing action is adjustable from a gentle slosh to a rapid agitation. It is ideal for use on a lab bench, in a small incubator or refrigerator, in a biological cabinet, or in a fume hood.
. For optimum mixing, the chemical resistant cabinet is fixed at a 20 degree angle. Unit comes complete with an aluminum disk head with spring clips to hold 15 each for both 5-10 mm and 12-14 mm tubes.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.