Learn More
Research Products International Corp Mini-Pipettor, 5ul, Red
SDP
Supplier:  Research Products International Corp 347022
Encompass_Preferred
Compact and engineered for precision dispensing, the sturdy Minipet air-displacement micropipettes come in a range of volumes (5ul to 200ul) and color options. Most important, the MiniPet micropipette provides as low as +/-3% accuracy and 2% precision at full stroke. Built from sturdy molded plastic, the air-displacement MiniPet micropipette handles thousands of pipettings.
Catalog No. 50-197-8537
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.